Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677850.3 | XP_005677907.1 | 1 - 2 | 257 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |