Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695224.3 | XP_005695281.1 | 4 - 5 | 1006 | 74 | |
GenBank mRNA | JO426510.1 | - | 4 - 5 | 1038 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |