Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059514.1 | XP_017915003.1 | 5 - 6 | 701 | 100 | |
GenBank mRNA | JO623762.2 | - | 5 - 6 | 716 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |