Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684818.3 | XP_005684875.3 | 4 - 5 | 1013 | 87 | |
GenBank mRNA | JO408772.2 | - | 3 - 4 | 408 | 87 | |
JO426190.1 | - | 5 - 6 | 1016 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |