Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681841.3 | XP_005681898.2 | 4 - 5 | 879 | 114 | |
XM_005681842.3 | XP_005681899.2 | 4 - 5 | 780 | 114 | ||
GenBank mRNA | JO594467.2 | - | 4 - 5 | 866 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |