Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685409.3 | XP_005685466.1 | 1 - 2 | 477 | 61 | |
XM_005685410.3 | XP_005685467.1 | 1 - 2 | 476 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |