Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686208.2 | XP_005686265.1 | 6 - 7 | 1309 | 89 | |
XM_005686209.2 | XP_005686266.1 | 5 - 6 | 832 | 89 | ||
XM_013967400.1 | XP_013822854.1 | 5 - 6 | 840 | 89 | ||
XM_013967401.1 | XP_013822855.1 | 5 - 6 | 789 | 89 | ||
XM_013967402.1 | XP_013822856.1 | 4 - 5 | 598 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |