Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973756.2 | XP_013829210.2 | 11 - 12 | 2780 | 82 | |
GenBank mRNA | KA343400.1 | - | 3 - 4 | 438 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |