Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048413.1 | XP_017903902.1 | 7 - 8 | 1543 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |