Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056115.1 | XP_017911604.1 | 4 - 5 | 541 | 58 | |
XM_018056116.1 | XP_017911605.1 | 3 - 4 | 464 | 58 | ||
XM_018056117.1 | XP_017911606.1 | 3 - 4 | 445 | 58 | ||
XM_018056118.1 | XP_017911607.1 | 2 - 3 | 329 | 58 | ||
XM_018056119.1 | XP_017911608.1 | 4 - 5 | 541 | 58 | ||
GenBank mRNA | JO346648.2 | - | 6 - 7 | 1794 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |