Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048847.1 | XP_017904336.1 | 3 - 4 | 5976 | 79 | |
XM_018048848.1 | XP_017904337.1 | 3 - 4 | 5976 | 79 | ||
XM_018048849.1 | XP_017904338.1 | 3 - 4 | 5976 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |