Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964434.2 | XP_013819888.2 | 5 - 6 | 764 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |