Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040552.1 | XP_017896041.1 | 6 - 7 | 1143 | 68 | |
XM_018040553.1 | XP_017896042.1 | 6 - 7 | 1140 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |