Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040766.1 | XP_017896255.1 | 7 - 8 | 788 | 112 | |
XM_018040767.1 | XP_017896256.1 | 7 - 8 | 874 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |