Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693877.3 | XP_005693934.1 | 3 - 4 | 626 | 83 | |
XM_005693878.3 | XP_005693935.2 | 3 - 4 | 454 | 83 | ||
GenBank mRNA | JO432881.2 | - | 3 - 4 | 666 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |