Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972780.2 | XP_013828234.1 | 2 - 3 | 286 | 100 | |
XM_018065559.1 | XP_017921048.1 | 2 - 3 | 286 | 100 | ||
XM_018065561.1 | XP_017921050.1 | 2 - 3 | 286 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |