Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065926.1 | XP_017921415.1 | 18 - 19 | 3751 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |