Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696602.3 | XP_005696659.2 | 3 - 4 | 489 | 97 | |
GenBank mRNA | JO324472.2 | - | 2 - 3 | 678 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |