Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055296.1 | XP_017910785.1 | 6 - 7 | 2473 | 54 | |
XM_018055297.1 | XP_017910786.1 | 6 - 7 | 2473 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |