Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041987.1 | XP_017897476.1 | 2 - 3 | 1539 | 106 | |
GenBank mRNA | JO344421.2 | - | 3 - 4 | 1035 | 106 | |
JP351143.2 | - | 2 - 3 | 875 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |