Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688174.3 | XP_005688231.2 | 2 - 3 | 406 | 64 | |
GenBank mRNA | JO631886.2 | - | 2 - 3 | 1593 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |