Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046967.1 | XP_017902456.1 | 2 - 3 | 851 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |