Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695308.3 | XP_005695365.1 | 2 - 3 | 407 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |