Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054120.1 | XP_017909609.1 | 2 - 3 | 636 | 83 | |
GenBank mRNA | JO410758.2 | - | 4 - 5 | 453 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |