Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065196.1 | XP_017920685.1 | 3 - 4 | 510 | 93 | |
GenBank mRNA | JO611799.2 | - | 2 - 3 | 728 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |