Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC108636028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048364.1 | XP_017903853.1 | 6 - 7 | 1107 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |