Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO519156.2 | - | 2 - 3 | 338 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |