Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062337.1 | XP_017917826.1 | 4 - 5 | 612 | 95 | |
XM_018062339.1 | XP_017917828.1 | 4 - 5 | 609 | 95 | ||
XM_018062340.1 | XP_017917829.1 | 4 - 5 | 612 | 95 | ||
XM_018062341.1 | XP_017917830.1 | 4 - 5 | 612 | 95 | ||
GenBank mRNA | JO449432.2 | - | 4 - 5 | 563 | 95 | |
JO515262.2 | - | 1 - 2 | 606 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |