Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064316.1 | XP_017919805.1 | 3 - 4 | 1211 | 104 | |
GenBank mRNA | JO440177.2 | - | 3 - 4 | 555 | 104 | |
KA347549.1 | - | 3 - 4 | 540 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |