Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063939.1 | XP_017919428.1 | 3 - 4 | 532 | 73 | |
XM_018063940.1 | XP_017919429.1 | 4 - 5 | 660 | 73 | ||
XM_018063941.1 | XP_017919430.1 | 3 - 4 | 715 | 73 | ||
GenBank mRNA | JO320460.2 | - | 3 - 4 | 726 | 73 | |
JO427631.1 | - | 4 - 5 | 73 | |||
JO428131.1 | - | 3 - 4 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |