Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049710.1 | XP_017905199.1 | 7 - 8 | 1005 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |