Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692176.3 | XP_005692233.1 | 3 - 4 | 721 | 95 | |
GenBank mRNA | JO613222.2 | - | 3 - 4 | 684 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |