Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058468.1 | XP_017913957.1 | 3 - 4 | 3337 | 96 | |
XM_018058469.1 | XP_017913958.1 | 3 - 4 | 3336 | 96 | ||
GenBank mRNA | JO617467.2 | - | 3 - 4 | 322 | 96 | |
JO617468.2 | - | 3 - 4 | 322 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |