Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692977.2 | XP_005693034.2 | 1 - 2 | 36 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |