Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693936.3 | XP_005693993.2 | 1 - 2 | 260 | 77 | |
GenBank mRNA | JO577482.1 | - | 1 - 2 | 256 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |