Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055757.1 | XP_017911246.1 | 3 - 4 | 546 | 130 | |
GenBank mRNA | JO510467.2 | - | 1 - 2 | 533 | 130 | |
JQ308784.1 | - | 3 - 4 | 476 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |