Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690643.3 | XP_005690700.2 | 1 - 2 | 713 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |