Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062497.1 | XP_017917986.1 | 4 - 5 | 766 | 68 | |
GenBank mRNA | KA332933.1 | - | 1 - 2 | 1200 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |