Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055021.1 | XP_017910510.1 | 1 - 2 | 289 | 113 | |
GenBank mRNA | KA335876.1 | - | 1 - 2 | 257 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |