Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675436.3 | XP_005675493.2 | 6 - 7 | 6423 | 93 | |
XM_018048958.1 | XP_017904447.1 | 7 - 8 | 6621 | 93 | ||
XM_018048968.1 | XP_017904457.1 | 8 - 9 | 6498 | 93 | ||
XM_018048977.1 | XP_017904466.1 | 7 - 8 | 6501 | 93 | ||
XM_018048993.1 | XP_017904482.1 | 7 - 8 | 6498 | 93 | ||
XM_018049001.1 | XP_017904490.1 | 6 - 7 | 6179 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |