Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686384.3 | XP_005686441.1 | 2 - 3 | 344 | 61 | |
XM_018055079.1 | XP_017910568.1 | 2 - 3 | 582 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |