Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040779.1 | XP_017896268.1 | 2 - 3 | 264 | 72 | |
GenBank mRNA | JO421992.2 | - | 2 - 3 | 265 | 72 |
Gene Symbol: PMP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO560005.2 | - | 1556 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |