Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063849.1 | XP_017919338.1 | 5 - 6 | 981 | 59 | |
GenBank mRNA | JO421557.2 | - | 1 - 2 | 1696 | 59 | |
KA317111.1 | - | 1 - 2 | 2478 | 59 | ||
KT315580.1 | - | 5 - 6 | 929 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |