Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055338.1 | XP_017910827.1 | 3 - 4 | 642 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |