Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102189642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038501.1 | XP_017893990.1 | 14 - 15 | 2134 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |