Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041630.1 | XP_017897119.1 | 1 - 2 | 1702 | 114 | |
XM_018041631.1 | XP_017897120.1 | 1 - 2 | 1706 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |