Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971523.2 | XP_013826977.2 | 1 - 2 | 1417 | 59 | |
GenBank mRNA | JO437124.2 | - | 1 - 2 | 168 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |