Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102180271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677291.3 | XP_005677348.2 | 1 - 2 | 652 | 81 | |
XM_018046343.1 | XP_017901832.1 | 1 - 2 | 652 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |