Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048643.1 | XP_017904132.1 | 7 - 8 | 2524 | 107 | |
XM_018048644.1 | XP_017904133.1 | 7 - 8 | 2524 | 107 | ||
XM_018048645.1 | XP_017904134.1 | 7 - 8 | 2524 | 107 | ||
XM_018048646.1 | XP_017904135.1 | 7 - 8 | 2524 | 107 | ||
XM_018048647.1 | XP_017904136.1 | 5 - 6 | 1648 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |