Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692322.3 | XP_005692379.2 | 3 - 4 | 292 | 56 | |
GenBank mRNA | JO645048.2 | - | 3 - 4 | 352 | 56 | |
JO645049.2 | - | 3 - 4 | 352 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |